1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR160511000
Price (MYR)
424.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Chemical Identifiers
CAS5965-83-3
IUPAC Name2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular FormulaC9H14O6S
InChI KeyNFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILESC.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.1 %
Salicylic acid=<0.04 % (HOC6H4COOH)
Insoluble matter=<0.02 %
Sulfate (SO4)=<0.02 %
Chloride (Cl)=<0.001 %