1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR160510025
Price (MYR)
2,196.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,196.00
Add to cart
Specifications
Chemical Name or Material1-Chloro-2, 4-dinitrobenzene
Name Note99%
CAS97-00-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger