2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR160200050
Price (MYR)
643.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
643.00
Add to cart
Chemical Identifiers
CAS116-11-0
IUPAC Name2-methoxyprop-1-ene
Molecular FormulaC4H8O
InChI KeyYOWQWFMSQCOSBA-UHFFFAOYSA-N
SMILESCOC(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=96.0 %
Stabilizer=<0.5 % potassium carbonate