5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
CAS: 1761-61-1 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR159911000
Price (MYR)
1,002.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,002.00
Add to cart
Specifications
Chemical Name or Material5-Bromosalicylaldehyde
Name Note99%
CAS1761-61-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning