5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
CAS: 1761-61-1 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR159910250
Price (MYR)
523.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
523.00
Add to cart
Chemical Identifiers
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 %
UVat 260 nm A: =<0.02
UV(0.5 M in water)
Loss on drying=<1 %
Appearance (Color)White