Dimethyl malonate, 97%, Thermo Scientific Chemicals
Dimethyl malonate, 97%, Thermo Scientific Chemicals
Dimethyl malonate, 97%, Thermo Scientific Chemicals
Dimethyl malonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl malonate, 97%, Thermo Scientific Chemicals
CAS: 108-59-8 | C5H8O4 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number ACR159560010
Price (MYR)
1,039.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,039.00
Add to cart
Chemical Identifiers
CAS1333-82-0
IUPAC Nametrioxochromium
Molecular FormulaCrO3
InChI KeyWGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILESO=[Cr](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.01%
Assay (unspecified)98.0% CrO₃ min
Impurity contentAl 0.02%
Impurity contentBa 0.01%
Impurity contentCl 0.005%