2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals
2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals
2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals
2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR159490250
Price (MYR)
680.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
680.00
Add to cart
Chemical Identifiers
CAS55934-00-4
IUPAC Name3,4-dichloropyridine
Molecular FormulaC5H3Cl2N
InChI KeyZMPYQKNNUHPTLT-UHFFFAOYSA-N
SMILESClC1=C(Cl)C=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%