Chalcone, 97%, Thermo Scientific Chemicals
Chalcone, 97%, Thermo Scientific Chemicals
Chalcone, 97%, Thermo Scientific Chemicals
Chalcone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chalcone, 97%, Thermo Scientific Chemicals
CAS: 94-41-7 | C15H12O | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR159431000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.80 %
Residue after evaporation=<0.0005 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Water=<0.02 % (K.F.)