4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
CAS: 74-11-3 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR159250250
Price (MYR)
385.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
385.00
Add to cart
Chemical Identifiers
CAS71519-72-7
IUPAC Namedisodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular FormulaC4H16N3Na2O9P
InChI KeyHUWYWJSJJDCZRQ-UHFFFAOYSA-L
SMILESO.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Heavy metals (as Pb)=<20 ppm
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Water21 to 25 % (K.F.)