2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2 kg
Packaging:
View all
Plastic bottle
Catalog number ACR159245000
Price (MYR)
534.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
534.00
Add to cart
Chemical Identifiers
CAS93-55-0
IUPAC Name1-phenylpropan-1-one
Molecular FormulaC9H10O
InChI KeyKRIOVPPHQSLHCZ-UHFFFAOYSA-N
SMILESCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥98.5%
Refractive Index1.5245-1.5280 @ 20?C