2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2 kg
Packaging:
View all
Plastic bottle
Catalog number ACR159241000
Price (MYR)
357.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
357.00
Add to cart
Specifications
Chemical Name or Material2-Chlorobenzoic acid
Name Note98%
CAS118-91-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning