2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
View all
Glass Bottle
Catalog number ACR159240050
Price (MYR)
366.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
366.00
Add to cart
Specifications
Chemical Name or Material2-Chlorobenzoic acid
Name Note98%
CAS118-91-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning