Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
CAS: 501-52-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number ACR159101000
Price (MYR)
513.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS108-99-6
IUPAC Name3-methylpyridine
Molecular FormulaC6H7N
InChI KeyITQTTZVARXURQS-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive index1.5050 to 1.5070 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %