Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrocinnamic acid, 99%, Thermo Scientific Chemicals
CAS: 501-52-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR159100050
Price (MYR)
397.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
397.00
Add to cart
Chemical Identifiers
CAS98-94-2
IUPAC NameN,N-dimethylcyclohexanamine
Molecular FormulaC8H17N
InChI KeySVYKKECYCPFKGB-UHFFFAOYSA-N
SMILESCN(C)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4525 to 1.4545 (20°C, 589 nm)
Color scale=<50 APHA