Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR158992500
Price (MYR)
498.00
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
498.00
Add to cart
Chemical Identifiers
CAS111-43-3
IUPAC Name1-propoxypropane
Molecular FormulaC6H14O
InChI KeyPOLCUAVZOMRGSN-UHFFFAOYSA-N
SMILESCCCOCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.3790-1.3820 @ 20?C