Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Packaging:
Glass bottle
Catalog number ACR158990010
Price (MYR)
993.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
993.00
Add to cart
Specifications
Chemical Name or MaterialThiazolyl blue tetrazolium bromide
Name Note98%
CAS298-93-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning