Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number ACR158922500
Price (MYR)
338.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
338.00
Add to cart
Specifications
Chemical Name or MaterialGlycerol
Name NoteExtra Pure
CAS56-81-5
Decomposition Information>290°C
Melting Point18.0°C