Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
CAS: 76-84-6 | C19H16O | 260.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number ACR158912500
Price (MYR)
1,170.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,170.00
Add to cart
Chemical Identifiers
CAS22398-80-7
IUPAC Nameindiganylidynephosphane
Molecular FormulaInP
InChI KeyGPXJNWSHGFTCBW-UHFFFAOYSA-N
SMILESP#[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to black
FormCrystalline solid
Total Metal Impurities< 100pm