Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethanol, 97%, Thermo Scientific Chemicals
CAS: 76-84-6 | C19H16O | 260.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number ACR158910500
Price (MYR)
496.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
496.00
Add to cart
Specifications
Chemical Name or MaterialTriphenylmethanol
CAS76-84-6
Melting Point160°C to 164°C
ColorWhite to Yellow
EINECS Number200-988-5