2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
CAS: 52-51-7 | C3H6BrNO4 | 199.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR158820250
Price (MYR)
466.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
466.00
Add to cart
Specifications
Chemical Name or Material2-Bromo-2-nitro-1, 3-propanediol
CAS52-51-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0