Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Succinic Acid 99%, Thermo Scientific Chemicals
CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR158742500
Price (MYR)
399.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
399.00
Add to cart
Chemical Identifiers
CAS618-89-3
IUPAC Namemethyl 3-bromobenzoate
Molecular FormulaC8H7BrO2
InChI KeyKMFJVYMFCAIRAN-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid or liquid
HPLC>=97.5 %
Appearance (Color)White to light yellow