trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
CAS: 140-10-3 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR158575000
Price (MYR)
747.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
747.00
Add to cart
Chemical Identifiers
CAS85117-99-3
IUPAC Name2-(bromomethyl)-1,4-difluorobenzene
Molecular FormulaC7H5BrF2
InChI KeyONWGSWNHQZYCFK-UHFFFAOYSA-N
SMILESFC1=CC=C(F)C(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5250-1.5290 @ 20?C
Assay (GC)≥97.5%