trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%, Thermo Scientific Chemicals
CAS: 140-10-3 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR158571000
Price (MYR)
402.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
402.00
Add to cart
Chemical Identifiers
CAS50-69-1
IUPAC Name(3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
Molecular FormulaC5H10O5
InChI KeyHMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILESOC[C@H]1OC(O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (60°C, 3 h)
Specific optical rotation-18° to -22° (20°C, 589 nm) (c=2, water)
Appearance (Color)White to light beige
Appearance (Form)Powder or crystalline powder