Chloroform, 99+%, Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR158210025
Price (MYR)
887.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
887.00
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content(Iodide, I) : 0.002% max
Impurity content(Iron, Fe) : 3ppm max
Impurity content(Magnesium, Mg) : 0.001% max
Impurity content(Phosphate, PO₄) : 5ppm max
Impurity content(Sodium, Na) : 0.005% max