Chloroform, 99+%, Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR158210010
Price (MYR)
563.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
563.00
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
CAS67-66-3
Health Hazard 1Warning
Health Hazard 2GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.