Thymine, 99%, Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thymine, 99%, Thermo Scientific Chemicals
CAS: 65-71-4 | C5H6N2O2 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR157851000
Price (MYR)
1,063.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,063.00
Add to cart
Chemical Identifiers
CAS97-72-3
IUPAC Name2-methylpropanoyl 2-methylpropanoate
Molecular FormulaC8H14O3
InChI KeyLSACYLWPPQLVSM-UHFFFAOYSA-N
SMILESCC(C)C(=O)OC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4040-1.4090 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%