4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
5 g
100 g
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR157050051
Price (MYR)
3,644.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
3,644.00
Add to cart
Chemical Identifiers
CAS13395-36-3
IUPAC Nameethyl 5,5-dimethyl-2,4-dioxohexanoate
Molecular FormulaC10H16O4
InChI KeyNIMKIMUBJFWPTD-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4675-1.4715 @ 20?C
Assay (GC)≥97.5%
Indicator
RUO – Research Use Only