4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
250 g
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number ACR157050050
Price (MYR)
498.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
498.00
Add to cart
Chemical Identifiers
CAS99-96-7
IUPAC Name4-hydroxybenzoic acid
Molecular FormulaC7H6O3
InChI KeyFJKROLUGYXJWQN-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay (Silylated GC)≥98.5% (non-U.S. sourced material)
Assay from Supplier's CofA≥98.5 (U.S. sourced material)
View more
Indicator
RUO – Research Use Only