4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenol, 99%, Thermo Scientific Chemicals
CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR157050010
Price (MYR)
1,144.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,144.00
Add to cart
Specifications
Chemical Name or Material4-Nitrophenol
CAS100-02-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful in contact with skin.
Toxic if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
View more
Indicator
RUO – Research Use Only