4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR156995000
Price (MYR)
3,043.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,043.00
Add to cart
Chemical Identifiers
CAS104219-63-8
SpecificationsSpecification SheetSpecification Sheet
Particle Size≤ 2.0% @ > 1.180mm
Appearance (Color)White to cream
FormBeads
Residual water62 - 70%
Particle Size≤ 2.0% @ < 0.300mm