4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR156991000
Price (MYR)
873.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
873.00
Add to cart
Chemical Identifiers
CAS4130-54-5
IUPAC Nameoctadecanehydrazide
Molecular FormulaC18H38N2O
InChI KeyBYTFESSQUGDMQQ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder
Melting Point (clear melt)104-111?C
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%