4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR156990250
Price (MYR)
513.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS811-98-3
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=99.75 %
Water=<0.03 %
Appearance (Form)Liquid