1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
CAS: 1445-73-4 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR156921000
Price (MYR)
724.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
724.00
Add to cart
Chemical Identifiers
CAS83-56-7
IUPAC Namenaphthalene-1,5-diol
Molecular FormulaC10H8O2
InChI KeyBOKGTLAJQHTOKE-UHFFFAOYSA-N
SMILESOC1=CC=CC2=C(O)C=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to brown to gray
Assay (HPLC)≥97.5%