2,4-Dihydroxybenzophenone 99%
2,4-Dihydroxybenzophenone 99%
2,4-Dihydroxybenzophenone 99%
2,4-Dihydroxybenzophenone 99%
2,4-Dihydroxybenzophenone 99%
CAS: 131-56-6 | C13H10O3 | 214.22 g/mol
Have Questions?
Catalog number ACR155181000
Price (MYR)
1.00
Add to cart
Request bulk or custom format
Price (MYR)
1.00
Add to cart
Chemical Identifiers
CAS2426-87-1
IUPAC Name4-(benzyloxy)-3-methoxybenzaldehyde
Molecular FormulaC15H14O3
InChI KeyJSHLOPGSDZTEGQ-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC=C1OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)59.0-66.0?C
Assay (GC)≥97.5%
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Free acid (titration)≤1%