Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR154960025
Price (MYR)
669.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
669.00
Add to cart
Chemical Identifiers
CAS5794-88-7
IUPAC Name2-amino-5-bromobenzoic acid
Molecular FormulaC7H6BrNO2
InChI KeyCUKXRHLWPSBCTI-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Powder
GC>=96.0 %