Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Glass bottle
Catalog number ACR154792500
Price (MYR)
1,120.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,120.00
Add to cart
Specifications
Chemical Name or MaterialCitraconic anhydride
Name Note98%
CAS616-02-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Toxic in contact with skin.