Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citraconic anhydride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Glass bottle
Catalog number ACR154791000
Price (MYR)
617.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
617.00
Add to cart
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH6.0-7.5 (1% solution)
UV/VisAbsorbance 280 nm (3% in water): ≤0.4
Chloride content≤0.10%
Solution TestPasses (10% in water)
Assay from Suppliers CofA≥99.0% (unspecified)