4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR154761000
Price (MYR)
595.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
595.00
Add to cart
Specifications
Chemical Name or Material4-Chlorothiophenol
Name Note98%
CAS106-54-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger