2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals
2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals
2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals
2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals
CAS: 2516-96-3 | C7H4ClNO4 | 201.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR154691000
Price (MYR)
502.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Specifications
Chemical Name or Material2-Chloro-5-nitrobenzoic acid
Name Note99+%
CAS2516-96-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning