Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Catalog number ACR154515000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Physical FormAdhering Crystals