Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR154511000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS67874-71-9
IUPAC Namebismuth(3+) tris(2-ethylhexanoate)
Molecular FormulaC24H45BiO6
InChI KeyNUMHJBONQMZPBW-UHFFFAOYNA-K
SMILES[Bi+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
FormViscous liquid
Assay from Supplier's CofA≥60%