Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR154510250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS882-33-7
IUPAC Name(phenyldisulfanyl)benzene
Molecular FormulaC12H10S2
InChI KeyGUUVPOWQJOLRAS-UHFFFAOYSA-N
SMILESS(SC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)57.0-63.0?C