4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR154355000
Price (MYR)
890.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
890.00
BOT
Add to cart
Chemical Identifiers
CAS458532-86-0
IUPAC Name2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular FormulaC11H15BFNO2
InChI KeyPCLMNCBIXQQRMB-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥94.0%
Melting Point (clear melt)44.5-53.5?C