4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR154351000
Price (MYR)
428.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
428.00
Add to cart
Specifications
Chemical Name or Material4-tert-Butylcatechol
Name Note99%
CAS98-29-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger