4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals
CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number ACR154350025
Price (MYR)
1,875.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,875.00
Add to cart
Chemical Identifiers
CAS15170-57-7
IUPAC Nameplatinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O4Pt
InChI KeyKLFRPGNCEJNEKU-FDGPNNRMSA-L
SMILES[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97.5 % (ex Pt)
Appearance (Form)Powder
Appearance (Color)Yellow to yellow-green