1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
Catalog number ACR154310025
Price (MYR)
8,354.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
8,354.00
Add to cart
Chemical Identifiers
CAS632-69-9
IUPAC Namedisodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular FormulaC20H2Cl4I4Na2O5
InChI KeyKCQREHTWEUECQT-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C(I)C=C2C(OC3=C(I)C([O-])=C(I)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test0.1 wt.% in water (+ 1ml 1% Na₂CO₃): Clear
lambda-max (UV-Vis)540-550nm (in water)
Specific Absorbance (E 1%/1cm)(at |l max) : ≥600
Appearance (Color)Red to dark red to brown