Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals
CAS: 873-55-2 | C6H5NaO2S | 164.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR154051000
Price (MYR)
593.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
593.00
Add to cart
Chemical Identifiers
CAS15931-15-4
IUPAC Name3-fluoro-2-methylpyridine
Molecular FormulaC6H6FN
InChI KeyIRIHSZDBTACXCT-UHFFFAOYSA-N
SMILESCC1=C(F)C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%