9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
CAS: 642-31-9 | C15H10O | 206.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR153931000
Price (MYR)
1,082.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,082.00
Add to cart
Specifications
Chemical Name or Material9-Anthracenecarboxaldehyde
CAS642-31-9
Melting Point103.0°C to 107.0°C
ColorYellow
EINECS Number211-383-