2-Aminothiophenol, 98%, Thermo Scientific Chemicals
2-Aminothiophenol, 98%, Thermo Scientific Chemicals
2-Aminothiophenol, 98%, Thermo Scientific Chemicals
2-Aminothiophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminothiophenol, 98%, Thermo Scientific Chemicals
CAS: 137-07-5 | C6H7NS | 125.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR153901000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Aminothiophenol
Name Note98%
CAS137-07-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger