3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Acros Organics, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR153850010
Price (MYR)
470.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
470.00
Add to cart
Chemical Identifiers
CAS7550-35-8
IUPAC Namelithium(1+) bromide
Molecular FormulaBrLi
InChI KeyAMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES[Li+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 %
Sulfate (SO4)=<200 ppm
Copper (Cu)=<3 ppm
Magnesium (Mg)=<20 ppm
Appearance (Color)White